Assay Details
| Gene Symbol: AT5G16070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121612.2 | NP_197111.2 | 4 - 5 | 618 | 70 | |
| GenBank mRNA | AY128362.1 | - | 4 - 5 | 618 | 70 | |
| BT008893.1 | - | 4 - 5 | 489 | 70 | ||
More Information
Set Membership: |
|

AT5G16070
Genomic Map