Assay Details
| Gene Symbol: SYN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121632.4 | NP_197131.2 | 4 - 5 | 811 | 76 | |
| GenBank mRNA | AF400129.1 | - | 4 - 5 | 709 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYN4| Gene Symbol: SYN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121632.4 | NP_197131.2 | 4 - 5 | 811 | 76 | |
| GenBank mRNA | AF400129.1 | - | 4 - 5 | 709 | 76 | |
Set Membership: |
|