Assay Details
| Gene Symbol: SYN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121632.4 | NP_197131.2 | 7 - 8 | 1008 | 106 | |
| GenBank mRNA | AF400129.1 | - | 7 - 8 | 906 | 106 | |
More Information
Set Membership: |
|

SYN4
Genomic Map