Assay Details
| Gene Symbol: UCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121636.3 | NP_197135.1 | 3 - 4 | 357 | 65 | |
| GenBank mRNA | AK118002.1 | - | 3 - 4 | 355 | 65 | |
| BT005312.1 | - | 3 - 4 | 225 | 65 | ||
More Information
Set Membership: |
|

UCH1
Genomic Map