Assay Details
| Gene Symbol: AT5G16590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121665.4 | NP_197162.1 | 1 - 2 | 1559 | 95 | |
| GenBank mRNA | AK228885.1 | - | 1 - 2 | 1462 | 95 | |
| BT011691.1 | - | 1 - 2 | 1381 | 95 | ||
| BT012270.1 | - | 1 - 2 | 1281 | 95 | ||
| FJ708778.1 | - | 1 - 2 | 1281 | 95 | ||
More Information
Set Membership: |
|

AT5G16590
Genomic Map