Assay Details
| Gene Symbol: AT5G16590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121665.4 | NP_197162.1 | 2 - 3 | 1885 | 72 | |
| GenBank mRNA | AK228885.1 | - | 2 - 3 | 1788 | 72 | |
| BT011691.1 | - | 2 - 3 | 1707 | 72 | ||
| BT012270.1 | - | 2 - 3 | 1607 | 72 | ||
| FJ708778.1 | - | 2 - 3 | 1607 | 72 | ||
More Information
Set Membership: |
|

AT5G16590
Genomic Map