Assay Details
| Gene Symbol: ITPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121682.3 | NP_197178.1 | 1 | 1538 | 86 | |
| GenBank mRNA | AY072156.1 | - | 1 | 1407 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITPK1| Gene Symbol: ITPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121682.3 | NP_197178.1 | 1 | 1538 | 86 | |
| GenBank mRNA | AY072156.1 | - | 1 | 1407 | 86 | |
Set Membership: |
|