Assay Details
| Gene Symbol: SYP21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121689.4 | NP_197185.1 | 3 - 4 | 579 | 111 | |
| GenBank mRNA | AF411797.1 | - | 3 - 4 | 497 | 111 | |
| BT025036.1 | - | 3 - 4 | 436 | 111 | ||
| L41651.1 | - | 3 - 4 | 540 | 111 | ||
More Information
Set Membership: |
|

SYP21
Genomic Map