Assay Details
| Gene Symbol: SYP21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121689.4 | NP_197185.1 | 4 - 5 | 661 | 105 | |
| GenBank mRNA | AF411797.1 | - | 4 - 5 | 579 | 105 | |
| BT025036.1 | - | 4 - 5 | 518 | 105 | ||
| L41651.1 | - | 4 - 5 | 622 | 105 | ||
More Information
Set Membership: |
|

SYP21
Genomic Map