Assay Details
| Gene Symbol: AT5G16900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343470.1 | NP_001318574.1 | 12 - 13 | 2131 | 150 | |
| NM_001343472.1 | NP_001331987.1 | 9 - 10 | 1594 | 150 | ||
| NM_121696.3 | NP_197192.2 | 11 - 12 | 2063 | 150 | ||
| GenBank mRNA | FJ708779.1 | - | 11 - 12 | 2038 | 150 | |
More Information
Set Membership: |
|

AT5G16900
Genomic Map