Assay Details
| Gene Symbol: AT5G16900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343470.1 | NP_001318574.1 | 3 - 4 | 698 | 134 | |
| NM_001343471.1 | NP_001331986.1 | 2 - 3 | 637 | 134 | ||
| NM_121696.3 | NP_197192.2 | 2 - 3 | 630 | 134 | ||
| GenBank mRNA | FJ708779.1 | - | 2 - 3 | 605 | 134 | |
More Information
Set Membership: |
|

AT5G16900
Genomic Map