Assay Details
| Gene Symbol: AT5G16900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343470.1 | NP_001318574.1 | 4 - 5 | 1200 | 84 | |
| NM_001343471.1 | NP_001331986.1 | 3 - 4 | 1139 | 84 | ||
| NM_001343472.1 | NP_001331987.1 | 1 - 2 | 663 | 84 | ||
| NM_121696.3 | NP_197192.2 | 3 - 4 | 1132 | 84 | ||
| GenBank mRNA | FJ708779.1 | - | 3 - 4 | 1107 | 84 | |
More Information
Set Membership: |
|

AT5G16900
Genomic Map