Assay Details
| Gene Symbol: AT5G16900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343470.1 | NP_001318574.1 | 6 - 7 | 1423 | 116 | |
| NM_001343471.1 | NP_001331986.1 | 5 - 6 | 1362 | 116 | ||
| NM_001343472.1 | NP_001331987.1 | 3 - 4 | 886 | 116 | ||
| NM_121696.3 | NP_197192.2 | 5 - 6 | 1355 | 116 | ||
| GenBank mRNA | FJ708779.1 | - | 5 - 6 | 1330 | 116 | |
More Information
Set Membership: |
|

AT5G16900
Genomic Map