Assay Details
| Gene Symbol: AT5G16900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343470.1 | NP_001318574.1 | 7 - 8 | 1487 | 79 | |
| NM_001343471.1 | NP_001331986.1 | 6 - 7 | 1426 | 79 | ||
| NM_001343472.1 | NP_001331987.1 | 4 - 5 | 950 | 79 | ||
| NM_121696.3 | NP_197192.2 | 6 - 7 | 1419 | 79 | ||
| GenBank mRNA | FJ708779.1 | - | 6 - 7 | 1394 | 79 | |
More Information
Set Membership: |
|

AT5G16900
Genomic Map