Assay Details
| Gene Symbol: AT5G16900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343470.1 | NP_001318574.1 | 8 - 9 | 1563 | 74 | |
| NM_001343471.1 | NP_001331986.1 | 7 - 8 | 1502 | 74 | ||
| NM_001343472.1 | NP_001331987.1 | 5 - 6 | 1026 | 74 | ||
| NM_121696.3 | NP_197192.2 | 7 - 8 | 1495 | 74 | ||
| GenBank mRNA | FJ708779.1 | - | 7 - 8 | 1470 | 74 | |
More Information
Set Membership: |
|

AT5G16900
Genomic Map