Assay Details
| Gene Symbol: XPO1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203395.1 | NP_001190324.1 | 12 - 13 | 1306 | 105 | |
| NM_121708.3 | NP_197204.1 | 13 - 14 | 1443 | 105 | ||
| GenBank mRNA | AY056126.1 | - | 13 - 14 | 1339 | 105 | |
| AY096633.1 | - | 13 - 14 | 1171 | 105 | ||
| Y18469.1 | - | 1266 | 105 | |||
More Information
Set Membership: |
|

XPO1A
Genomic Map