Assay Details
| Gene Symbol: XPO1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203395.1 | NP_001190324.1 | 3 - 4 | 488 | 105 | |
| NM_121708.3 | NP_197204.1 | 3 - 4 | 580 | 105 | ||
| GenBank mRNA | AY056126.1 | - | 3 - 4 | 476 | 105 | |
| AY096633.1 | - | 3 - 4 | 308 | 105 | ||
| Y18469.1 | - | 403 | 105 | |||
More Information
Set Membership: |
|

XPO1A
Genomic Map