Assay Details
| Gene Symbol: XPO1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203395.1 | NP_001190324.1 | 7 - 8 | 891 | 109 | |
| NM_121708.3 | NP_197204.1 | 7 - 8 | 983 | 109 | ||
| GenBank mRNA | AY056126.1 | - | 7 - 8 | 879 | 109 | |
| AY096633.1 | - | 7 - 8 | 711 | 109 | ||
| Y18469.1 | - | 806 | 109 | |||
More Information
Set Membership: |
|

XPO1A
Genomic Map