Assay Details
| Gene Symbol: ENH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085129.1 | NP_001078598.1 | 1 - 2 | 249 | 72 | |
| NM_121723.5 | NP_568342.1 | 1 - 2 | 385 | 72 | ||
| GenBank mRNA | AY035119.1 | - | 1 - 2 | 192 | 72 | |
| BT000984.1 | - | 1 - 2 | 107 | 72 | ||
More Information
Set Membership: |
|

ENH1
Genomic Map