Assay Details
| Gene Symbol: ENH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085129.1 | NP_001078598.1 | 5 - 6 | 783 | 136 | |
| NM_121723.5 | NP_568342.1 | 5 - 6 | 920 | 137 | ||
| GenBank mRNA | AY035119.1 | - | 5 - 6 | 727 | 137 | |
| BT000984.1 | - | 5 - 6 | 642 | 137 | ||
More Information
Set Membership: |
|

ENH1
Genomic Map