Assay Details
| Gene Symbol: APG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121735.5 | NP_197231.1 | 7 - 8 | 873 | 68 | |
| GenBank mRNA | AK117572.1 | - | 7 - 8 | 759 | 68 | |
| BT005100.1 | - | 7 - 8 | 747 | 68 | ||
More Information
Set Membership: |
|

APG5
Genomic Map