Assay Details
| Gene Symbol: GAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121739.4 | NP_197235.1 | 2 - 3 | 518 | 122 | |
| GenBank mRNA | AY094464.1 | - | 2 - 3 | 360 | 122 | |
| BT001047.1 | - | 2 - 3 | 287 | 122 | ||
| Gene Symbol: AT1G48740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230255.1 | - | 986 | 122 | ||
More Information
Set Membership: |
|

GAD
Genomic Map