Assay Details
| Gene Symbol: GAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121739.4 | NP_197235.1 | 7 - 8 | 1420 | 66 | |
| GenBank mRNA | AY094464.1 | - | 7 - 8 | 1262 | 66 | |
| BT001047.1 | - | 7 - 8 | 1189 | 66 | ||
| Gene Symbol: AT1G48740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230255.1 | - | 84 | 66 | ||
More Information
Set Membership: |
|

GAD
Genomic Map