Assay Details
| Gene Symbol: RCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121758.4 | NP_568349.1 | 1 - 2 | 453 | 82 | |
| GenBank mRNA | AF168390.1 | - | 1 - 2 | 285 | 82 | |
| AY056187.1 | - | 1 - 2 | 329 | 82 | ||
| AY096715.1 | - | 1 - 2 | 267 | 82 | ||
| Gene Symbol: AT5G17523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125768.3 | NP_001119240.2 | 1 - 2 | 386 | 82 | |
More Information
Set Membership: |
|

RCP1
Genomic Map