Assay Details
| Gene Symbol: RCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343506.1 | NP_001330869.1 | 3 - 4 | 1104 | 88 | |
| NM_121758.4 | NP_568349.1 | 4 - 5 | 1004 | 88 | ||
| GenBank mRNA | AF168390.1 | - | 4 - 5 | 836 | 88 | |
| AY056187.1 | - | 4 - 5 | 880 | 88 | ||
| AY096715.1 | - | 4 - 5 | 818 | 88 | ||
More Information
Set Membership: |
|

RCP1
Genomic Map