Assay Details
| Gene Symbol: RCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343506.1 | NP_001330869.1 | 6 - 7 | 1390 | 90 | |
| NM_121758.4 | NP_568349.1 | 7 - 8 | 1290 | 90 | ||
| GenBank mRNA | AF168390.1 | - | 7 - 8 | 1122 | 90 | |
| AY056187.1 | - | 7 - 8 | 1166 | 90 | ||
| AY096715.1 | - | 7 - 8 | 1104 | 90 | ||
More Information
Set Membership: |
|

RCP1
Genomic Map