Assay Details
| Gene Symbol: PEX19-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121761.3 | NP_568351.1 | 2 - 3 | 226 | 104 | |
| GenBank mRNA | AF410297.1 | - | 2 - 3 | 161 | 104 | |
| AY149931.1 | - | 2 - 3 | 88 | 104 | ||
More Information
Set Membership: |
|

PEX19-2
Genomic Map