Assay Details
| Gene Symbol: PEX19-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343512.1 | NP_001332000.1 | 3 - 4 | 944 | 62 | |
| NM_121761.3 | NP_568351.1 | 4 - 5 | 830 | 62 | ||
| GenBank mRNA | AF410297.1 | - | 4 - 5 | 765 | 62 | |
| AY149931.1 | - | 4 - 5 | 692 | 62 | ||
More Information
Set Membership: |
|

PEX19-2
Genomic Map