Assay Details
| Gene Symbol: AT5G17560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343513.1 | NP_001330623.1 | 1 - 2 | 438 | 92 | |
| NM_001343514.1 | NP_001330622.1 | 1 - 2 | 438 | 92 | ||
| NM_121762.3 | NP_197258.1 | 1 - 2 | 438 | 92 | ||
| GenBank mRNA | AY088829.1 | - | 1 - 2 | 342 | 92 | |
| BT025531.1 | - | 1 - 2 | 322 | 92 | ||
More Information
Set Membership: |
|

AT5G17560
Genomic Map