Assay Details
| Gene Symbol: CAX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343527.1 | NP_001332508.1 | 2 | 1635 | 129 | |
| NM_121792.4 | NP_197288.1 | 1 | 1892 | 129 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAX7| Gene Symbol: CAX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343527.1 | NP_001332508.1 | 2 | 1635 | 129 | |
| NM_121792.4 | NP_197288.1 | 1 | 1892 | 129 | ||
Set Membership: |
|