Assay Details
| Gene Symbol: CSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343529.1 | NP_001331999.1 | 3 - 4 | 2189 | 107 | |
| NM_121794.2 | NP_197290.1 | 3 - 4 | 1971 | 107 | ||
More Information
Set Membership: |
|

CSA1
Genomic Map