Assay Details
| Gene Symbol: ATMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085130.2 | NP_001078599.1 | 4 - 5 | 795 | 93 | |
| NM_121798.4 | NP_197294.1 | 4 - 5 | 749 | 93 | ||
| GenBank mRNA | AF370522.1 | - | 4 - 5 | 650 | 93 | |
| AY048201.1 | - | 4 - 5 | 650 | 93 | ||
| AY056098.1 | - | 4 - 5 | 650 | 93 | ||
| AY057499.1 | - | 4 - 5 | 650 | 93 | ||
| AY069876.1 | - | 4 - 5 | 650 | 93 | ||
| AY070771.1 | - | 4 - 5 | 650 | 93 | ||
| AY091692.1 | - | 3 - 4 | 568 | 93 | ||
| U97200.1 | - | 3 - 4 | 576 | 93 | ||
More Information
Set Membership: |
|

ATMS1
Genomic Map