Assay Details
| Gene Symbol: PIR121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121846.4 | NP_197342.3 | 28 - 29 | 3392 | 68 | |
| NM_203072.3 | NP_974801.2 | 28 - 29 | 3513 | 68 | ||
| GenBank mRNA | AY496701.1 | - | 28 - 29 | 3440 | 68 | |
| AY662957.1 | - | 27 - 28 | 3266 | 68 | ||
| AY787212.1 | - | 27 - 28 | 3269 | 68 | ||
More Information
Set Membership: |
|

PIR121
Genomic Map