Assay Details
| Gene Symbol: EMB269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121862.3 | NP_197358.2 | 2 - 3 | 1502 | 83 | |
| GenBank mRNA | AY128395.1 | - | 2 - 3 | 1393 | 83 | |
| BT008421.1 | - | 2 - 3 | 1381 | 83 | ||
More Information
Set Membership: |
|

EMB269
Genomic Map