Assay Details
| Gene Symbol: BMY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121872.3 | NP_197368.1 | 1 - 2 | 431 | 74 | |
| GenBank mRNA | AY069879.1 | - | 1 - 2 | 432 | 74 | |
| AY087036.1 | - | 1 - 2 | 426 | 74 | ||
| AY142007.1 | - | 1 - 2 | 249 | 74 | ||
More Information
Set Membership: |
|

BMY3
Genomic Map