Assay Details
| Gene Symbol: AT5G18700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121875.4 | NP_197371.2 | 7 - 8 | 652 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G18700| Gene Symbol: AT5G18700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121875.4 | NP_197371.2 | 7 - 8 | 652 | 126 | |
Set Membership: |
|