Assay Details
| Gene Symbol: SCL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343570.1 | NP_001331795.1 | 2 - 3 | 421 | 95 | |
| NM_121886.4 | NP_197382.3 | 2 - 3 | 421 | 95 | ||
| GenBank mRNA | DQ446966.1 | - | 2 - 3 | 254 | 95 | |
More Information
Set Membership: |
|

SCL28
Genomic Map