Assay Details
| Gene Symbol: SCL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343570.1 | NP_001331795.1 | 5 - 6 | 682 | 78 | |
| NM_121886.4 | NP_197382.3 | 5 - 6 | 682 | 78 | ||
| GenBank mRNA | DQ446966.1 | - | 5 - 6 | 515 | 78 | |
More Information
Set Membership: |
|

SCL28
Genomic Map