Assay Details
| Gene Symbol: LUL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121913.3 | NP_197409.1 | 2 - 3 | 1270 | 59 | |
| GenBank mRNA | BT006357.1 | - | 2 - 3 | 1019 | 59 | |
More Information
Set Membership: |
|

LUL3
Genomic Map