Assay Details
| Gene Symbol: AT5G19370 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121942.5 | NP_568372.1 | 6 - 7 | 878 | 72 | |
| GenBank mRNA | AF370165.1 | - | 6 - 7 | 792 | 72 | |
| AY059127.1 | - | 6 - 7 | 772 | 72 | ||
More Information
Set Membership: |
|

AT5G19370
Genomic Map