Assay Details
| Gene Symbol: ROPGEF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343611.1 | NP_001318601.1 | 6 - 7 | 941 | 91 | |
| NM_001343612.1 | NP_001331396.1 | 5 - 6 | 1136 | 91 | ||
| NM_001343613.1 | NP_001331395.1 | 5 - 6 | 1152 | 91 | ||
| NM_001343614.1 | NP_001331397.1 | 5 - 6 | 1136 | 91 | ||
| NM_121961.2 | NP_197457.1 | 5 - 6 | 755 | 91 | ||
| GenBank mRNA | DQ459196.1 | - | 5 - 6 | 755 | 91 | |
More Information
Set Membership: |
|

ROPGEF10
Genomic Map