Assay Details
| Gene Symbol: ASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124949.1 | NP_001118421.1 | 5 - 6 | 687 | 77 | |
| NM_128651.5 | NP_180654.1 | 4 - 5 | 776 | 77 | ||
| GenBank mRNA | AY059912.1 | - | 4 - 5 | 644 | 77 | |
| AY128806.1 | - | 4 - 5 | 593 | 77 | ||
| U15026.1 | - | 4 - 5 | 691 | 77 | ||
More Information
Set Membership: |
|

ASP1
Genomic Map