Assay Details
| Gene Symbol: ASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124949.1 | NP_001118421.1 | 6 - 7 | 784 | 78 | |
| NM_128651.5 | NP_180654.1 | 5 - 6 | 873 | 78 | ||
| GenBank mRNA | AY059912.1 | - | 5 - 6 | 741 | 78 | |
| AY128806.1 | - | 5 - 6 | 690 | 78 | ||
| U15026.1 | - | 5 - 6 | 788 | 78 | ||
More Information
Set Membership: |
|

ASP1
Genomic Map