Assay Details
| Gene Symbol: ASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124949.1 | NP_001118421.1 | 7 - 8 | 960 | 132 | |
| NM_128651.5 | NP_180654.1 | 6 - 7 | 1049 | 132 | ||
| GenBank mRNA | AY059912.1 | - | 6 - 7 | 917 | 132 | |
| AY128806.1 | - | 6 - 7 | 866 | 132 | ||
| U15026.1 | - | 6 - 7 | 964 | 132 | ||
More Information
Set Membership: |
|

ASP1
Genomic Map