Assay Details
| Gene Symbol: SKdZeta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128652.4 | NP_180655.1 | 1 - 2 | 215 | 79 | |
| GenBank mRNA | AY064020.1 | - | 1 - 2 | 165 | 79 | |
| AY094423.1 | - | 1 - 2 | 191 | 79 | ||
| AY096698.1 | - | 1 - 2 | 112 | 79 | ||
More Information
Set Membership: |
|

SKdZeta
Genomic Map