Assay Details
| Gene Symbol: emb1381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128691.4 | NP_180692.3 | 10 - 11 | 1683 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

emb1381| Gene Symbol: emb1381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128691.4 | NP_180692.3 | 10 - 11 | 1683 | 96 | |
Set Membership: |
|