Assay Details
| Gene Symbol: AT2G31390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128696.4 | NP_180697.1 | 2 - 3 | 1002 | 76 | |
| GenBank mRNA | AY074365.1 | - | 2 - 3 | 814 | 76 | |
| AY085748.1 | - | 2 - 3 | 821 | 76 | ||
| AY091312.1 | - | 2 - 3 | 726 | 76 | ||
More Information
Set Membership: |
|

AT2G31390
Genomic Map