Assay Details
| Gene Symbol: SAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128723.3 | NP_180724.2 | 10 - 11 | 1676 | 151 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAD2| Gene Symbol: SAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128723.3 | NP_180724.2 | 10 - 11 | 1676 | 151 | |
Set Membership: |
|