Assay Details
| Gene Symbol: TEJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128745.3 | NP_565730.1 | 8 - 9 | 1311 | 132 | |
| GenBank mRNA | AF394690.1 | - | 8 - 9 | 1224 | 132 | |
More Information
Set Membership: |
|

TEJ
Genomic Map