Assay Details
| Gene Symbol: TEJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084520.1 | NP_001077989.1 | 9 - 10 | 1512 | 72 | |
| NM_128745.3 | NP_565730.1 | 9 - 10 | 1514 | 72 | ||
| GenBank mRNA | AF394690.1 | - | 9 - 10 | 1427 | 72 | |
More Information
Set Membership: |
|

TEJ
Genomic Map